Skip to Content
Merck
All Photos(2)

Key Documents

G9513

Sigma-Aldrich

γ-Globulins from goat

≥99% (agarose gel electrophoresis)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€460.00

€460.00


Check Cart for Availability
Diagnostics Manufacturers should use alternative grade product: SRE0011

Request a Bulk Order

Select a Size

Change View
1 G
€460.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

€460.00


Check Cart for Availability
Diagnostics Manufacturers should use alternative grade product: SRE0011

Request a Bulk Order

biological source

goat

Quality Level

Assay

≥99% (agarose gel electrophoresis)

form

powder

impurities

2-4% NaCl

storage temp.

−20°C

General description

γ-Globulins are glycoproteins that are antibodies produced by plasma cells. They are immunoglobulins and exist in five classes, namely: IgG, IgM, IgA, IgD, and IgE. Immunoglobulins are heterogeneous molecules that constitute 20% of the plasma proteins. They are made up of four polypeptide chains (two light chains and two heavy chains) linked together by disulfide bonds. The antigen-binding site is present at the nitrogen terminal.

Biochem/physiol Actions

Immunoglobulins are generated in response to antigens, and they serve as key regulators of the immune system. They protect our bodies against bacteria, viruses, and fungi. Immunoglobulins are involved in the activation of the complement system, opsonization of microbes for phagocytosis, neutralization of toxins and viruses, and inhibition of the attachment of microbes to mucosal surfaces. Immunodeficiency can lead to recurrent infectious diseases.

Preparation Note

Prepared from Cohn Fraction II, III

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Postoperative thrombosis prevention using low-dose heparin].
A E Schaer et al.
Archiv fur Gynakologie, 219(1-4), 545-545 (1975-11-18)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service