Skip to Content
Merck
All Photos(1)

Key Documents

G8919

Sigma-Aldrich

Anti-Growth Hormone Receptor antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-GHR

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€485.00

€485.00


Estimated to ship on15 May 2025



Select a Size

Change View
100 μG
€485.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€485.00


Estimated to ship on15 May 2025


biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

immunohistochemistry: 5-15 μg/mL using cells and tissues
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Ghr(14600)

General description

By immunoblotting, this antibody shows ~50% cross-reactivity with recombinant rat GHR and 20% cross-reactivty with recombinant human GHR.
Growth Hormone Receptor encodes a protein that is a transmembrane receptor for growth hormone. Binding of growth hormone to the receptor leads to receptor dimerization and the activation of intra- and intercellular signal transduction pathways.

Immunogen

purified, NSO-derived, recombinant mouse growth hormone receptor extracellular domain.

Application

Anti-Growth Hormone Receptor antibody produced in goat was used to study the protein and protein phosphorylation levels in rat primary hippocampal cultures using Li-cor Odyssey fluorescence technology.

Biochem/physiol Actions

Growth hormone (GH) is secreted by the anterior pituitary and functions by binding specific growth hormone receptor (GHR). GH stimulates the secretion of insulin-like growth factor-1 that mediates some of its growth promoting actions. The activity of GH/GHR is important for normal physiology and aging processes.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Ding et al.
Proteomics, 13(3-4), 674-685 (2012-09-29)
Growth hormone (GH) is a protein secreted by the anterior pituitary and circulates throughout the body to exert important actions on growth and metabolism. GH stimulates the secretion of insulin-like growth factor-I (IGF-I) that mediates some of the growth promoting
Zhongbo Liu et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 30(2), 635-652 (2015-10-21)
Bone minerals are acquired during growth and are key determinants of adult skeletal health. During puberty, the serum levels of growth hormone (GH) and its downstream effector IGF-1 increase and play critical roles in bone acquisition. The goal of the
George A Garinis et al.
Nature cell biology, 11(5), 604-615 (2009-04-14)
The accumulation of stochastic DNA damage throughout an organism's lifespan is thought to contribute to ageing. Conversely, ageing seems to be phenotypically reproducible and regulated through genetic pathways such as the insulin-like growth factor-1 (IGF-1) and growth hormone (GH) receptors
Andrzej Bartke et al.
Endokrynologia Polska, 64(1), 46-52 (2013-03-02)
Studies in mutant, gene knock-out and transgenic mice have demonstrated that growth hormone (GH) signalling has a major impact on ageing and longevity. Growth hormone-resistant and GH-deficient animals live much longer than their normal siblings, while transgenic mice overexpressing GH
F J Steyn et al.
Endocrinology, 154(12), 4695-4706 (2013-10-11)
GH deficiency is thought to be involved in the pathogenesis of amyotrophic lateral sclerosis (ALS). However, therapy with GH and/or IGF-I has not shown benefit. To gain a better understanding of the role of GH secretion in ALS pathogenesis, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service