Skip to Content
Merck
All Photos(3)

Key Documents

G8885

Sigma-Aldrich

β-Glucuronidase from Helix pomatia

Type H-3, aqueous solution, ≥90,000 units/mL

Synonym(s):

β-D-Glucuronide glucuronosohydrolase

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€53.00
10 ML
€265.00
25 ML
€525.00

€53.00


Check Cart for Availability


Select a Size

Change View
1 ML
€53.00
10 ML
€265.00
25 ML
€525.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

€53.00


Check Cart for Availability

type

Type H-3

Quality Level

form

aqueous solution

specific activity

≥90,000 units/mL

secondary activity

≤1,000 units/mL sulfatase

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

β-Glucuronidase Type H-2 from Helix pomatia is a crude solution of enzymes derived from the Roman snail. Many β-glucuronidases derived from mollusks also contain sulfatase activity.

Application

β-Glucuronidase from Helix pomatia has been used:
  • to convert isoflavone conjugates to their aglycone forms in urine and soymilk samples for high performance liquid chromatography (HPLC) detection[1]
  • as a component in the protoplasting solution[2]
  • for the generation of aromatic and phenolic compounds from the plasma and urine samples for gas chromatography-mass spectrometry (GC-MS)[3]
  • to hydrolyze glucuronide and sulfate conjugated metabolites in fecal samples[4]

β-glucuronidase was used in the measurement of aromatization by a urine technique suitable for the evaluation of aromatase inhibitors in vivo. [5]
New Technical Article Comparing Performance of Different Enzymes
Learn more
about recent application data generated by Sigma R&D to optimize hydrolysis for different drug classes using enzymes from different sources and the use of a chromatographicaly purified enzyme to reduce the effect of esterase activity resulting in conversion of 6-MAM to Morphine

Biochem/physiol Actions

β-glucuronidase (β-GIc) is an exoglycosidase that catalyzes the breakdown of complex carbohydrates. In humans it converts conjugated bilirubin into the unconjugated form, making bilirubin suitable for reabsorption.
Used for the hydrolysis of glucuronide conjugates in urinary metabolite analysis

Quality

Many β-glucuronidases derived from mollusks also contain sulfatase activity.

Unit Definition

One Sigma or modified Fishman unit will liberate 1.0 μg of phenolphthalein from phenolphthalein glucuronide per hr at 37 °C at pH 5.0 (30 min assay).
Sulfatase Unit Definition: One unit of sulfatase will hydrolyze 1.0 μmole p-nitrocatechol sulfate per hr at pH 5.0 at 37 °C.

Physical form

Aqueous solution in ~1.0 M ammonium sulfate with 3 mM sodium azide as preservative.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

β-glucuronidase (GUS) enzymes are utilized to hydrolyze glucuronide (gluc) drug metabolites to the parent drug, facilitating analysis by LC-MS/MS.

Protocols

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service