Skip to Content
Merck
All Photos(1)

Key Documents

G50300

Sigma-Aldrich

Sephadex® G-50

Coarse

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817

swelling

-1 g swells to 9-11 mL

bead size

100-300 μm

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sephadex® G-50 is a gel filtration medium used in affinity chromatography, protein chromatrography and gel filtration chromatography. Sephadex® G-50 has been used to investigate effects from snake venom, isolate lectins from various species and to demonstrate the positive influence of 20% ethanol on proteinase desorption.

Legal Information

Sephadex is a registered trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Shelub et al.
Journal of applied physiology: respiratory, environmental and exercise physiology, 56(3), 810-815 (1984-03-01)
Despite numerous efforts, a reliable model of chronic embolic pulmonary hypertension has not been established. To develop such a model five conscious mongrel dogs were embolized repeatedly over 16-30 wk with Sephadex microspheres 286 +/- 70 micron in diameter. Hemodynamic
Martin Jaburek et al.
The Journal of biological chemistry, 279(51), 53097-53102 (2004-10-12)
Functional activation of mitochondrial uncoupling protein-2 (UCP2) is proposed to decrease reactive oxygen species production. Skulachev and Goglia (Skulachev, V. P., and Goglia, F. (2003) FASEB J. 17, 1585-1591) hypothesized that hydroperoxy fatty acid anions are translocated by UCPs but
Li-Feng Huang et al.
Toxicon : official journal of the International Society on Toxinology, 51(6), 1008-1016 (2008-02-19)
A group of small proteins, designated as muscarinic toxins (MTs), have been isolated from the venom of African green mamba (Dendroaspis angusticeps) and documented to bind selectively to individual muscarinic acetylcholine receptor (mAChR) subtypes. These components have less been reported
Tomohiro Suzuki et al.
Glycobiology, 22(5), 616-629 (2011-12-27)
A lectin was purified from the mushroom Hygrophorus russula by affinity chromatography on a Sephadex G-50 column and BioAssist S cation exchange chromatography and designated H. russula lectin (HRL). The results of sodium dodecyl sulfate-polyaclylamidegel electrophoresis, gel filtration and matrix-assisted
Katharina Besemer et al.
Applied and environmental microbiology, 75(22), 7189-7195 (2009-09-22)
Streams are highly heterogeneous ecosystems, in terms of both geomorphology and hydrodynamics. While flow is recognized to shape the physical architecture of benthic biofilms, we do not yet understand what drives community assembly and biodiversity of benthic biofilms in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service