Skip to Content
Merck
All Photos(2)

Key Documents

G4776

Grids for transmission electron microscopy

grid size 200 mesh × 125 μm pitch, copper

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€38.10

€38.10


Estimated to ship on28 May 2025



Select a Size

Change View
1 VIAL
€38.10

About This Item

UNSPSC Code:
41100000
NACRES:
MA.01

€38.10


Estimated to ship on28 May 2025


material

copper

description

Regular grids with center mark and solid border

packaging

vial of 100 grids

diam.

3.05 mm

grid size

200 mesh × 125 μm pitch

hole W

97 μm

open area size

55%

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Grids for transmission electron microscopy has been used to collect ultrafine sections of murine macrophage cell line during transmission electron microscopy (TEM).

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanessa Cohignac et al.
Autophagy, 14(8), 1323-1334 (2018-06-26)
Nanoparticles (NPs) can be toxic, depending on their physico-chemical characteristics. Macroautophagy/autophagy could represent a potential underlying mechanism of this toxicity. We therefore set up a study aimed to characterize in depth the effects, on autophagy, of macrophage exposure to NPs

Questions

  1. Hello there, I'd like to buy some TEM copper grids with markers around. The grids (G4776) are shown to have letters and numbers in the picture, but the description says it's regular grids with only center mark. Could you please clarify?

    1 answer
    1. The Grids has a center mark. There's no numbers and letters.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service