Skip to Content
Merck
All Photos(1)

Key Documents

G211150

Sigma-Aldrich

DMT-2′O-Methyl-rG(ib) Phosphoramidite

configured for (ÄKTA® and OligoPilot®)

Synonym(s):

DMT-2′-O-Me-rG(ib) amidite, N-(2-methyl-1-oxopropyl)-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-methyl-guanosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€424.00
10 G
€849.00

€424.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€424.00
10 G
€849.00

About This Item

Empirical Formula (Hill Notation):
C45H56N7O9P
CAS Number:
Molecular Weight:
869.94
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.51

€424.00


Check Cart for Availability

Request a Bulk Order

biological source

non-animal source (no BSE/TSE risk)

Quality Level

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder

technique(s)

oligo synthesis: suitable

impurities

≤0.1% single unspecified Impurity (reversed phase HPLC)
≤0.3% mG2 (reversed phase HPLC, Hydrolysate)
≤0.3% mG3 (reversed phase HPLC, DMT-rG(ib)me)
≤0.3% water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤1.0% mG1 (reversed phase HPLC, DMT-rG(ib)me-DMT)
≤3% residual Solvent content

color

white to off-white

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

compatibility

configured for (ÄKTA® and OligoPilot®)

nucleoside profile

base: guanosine
base protecting group: isobutyryl
2' protecting group: methyl
5' protecting group: DMT
deprotection: standard

storage temp.

2-8°C

SMILES string

CO[C@@H]1[C@H](OP(OCCC#N)N(C(C)C)C(C)C)[C@@H](COC(c2ccccc2)(c3ccc(OC)cc3)c4ccc(OC)cc4)O[C@H]1n5cnc6C(=O)NC(NC(=O)C(C)C)=Nc56

InChI

1S/C45H56N7O9P/c1-28(2)41(53)49-44-48-40-37(42(54)50-44)47-27-51(40)43-39(57-9)38(61-62(59-25-13-24-46)52(29(3)4)30(5)6)36(60-43)26-58-45(31-14-11-10-12-15-31,32-16-20-34(55-7)21-17-32)33-18-22-35(56-8)23-19-33/h10-12,14-23,27-30,36,38-39,43H,13,25-26H2,1-9H3,(H2,48,49,50,53,54)/t36-,38-,39-,43-,62?/m1/s1

InChI key

IRRDHRZUOZNWDJ-MLLDKZSOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Proligo′s 2′O-Methyl RNA monomers are compatible with fast deprotectionschemes that are based on the application of aliphatic amines, such asmethylamine. The adenosine and guanosine monomers are protected bythe standard benzoyl (bz) and isobutyryl (ib) groups.2′O-Methyl RNA is a nucleic acid analog that is characterized by theexceptional hybridization properties that it imparts with complimentaryDNA or RNA, as well as increased stability against enzymatic degradationcompared to natural nucleic acids.The unique combination of properties of 2′O-Methyl RNA had foundwidespread use in the fields of:
  • Diagnostic probes
  • Aptamer and ribozyme development
  • Mixed 2′O-Methyl-RNA/DNA antisense molecules
Key Features:
  • High yield of crude oligonucleotides
  • Compatible with DNA synthesis
  • Can be employed together with DNA or RNA phosphoramidites in thesame synthesis to produce mixmer oligonucleotides
  • Recommended deprotection conditions are 8 hours at 55 °C usingconcentrated ammonia solution, or with AMA (concentrated ammonia/40% aqueous methylamine I/I, v/v) for 10 minutes at 65 °C
  • Purification and other downstream processing of fully modified 2′OMethyl RNA oligonucleotides are simpler than in the case of RNA, as nospecial precautions are required to provide protection against nucleolyticdegradationDMT-2′O-Methyl-rG(ib)
Phosphoramidite is configured for ÄKTA® Synthesizers.

Legal Information

OligoPilot is a registered trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service