Skip to Content
Merck
All Photos(1)

Key Documents

G211140

Sigma-Aldrich

DMT-2′O-Methyl-rG(ib) Phosphoramidite

configured for MerMade

Synonym(s):

DMT-2′-O-Me-rG(ib) amidite, N-(2-methyl-1-oxopropyl)-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-methyl-guanosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H56N7O9P
CAS Number:
Molecular Weight:
869.94
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.51
Pricing and availability is not currently available.

biological source

non-animal source (no BSE/TSE risk)

Quality Level

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder

mol wt

869.94 g/mol

impurities

≤0.3% mG2 (reversed phase HPLC, Hydrolysate)
≤0.3% mG3 (reversed phase HPLC, DMT-rG(ib)me)
≤0.3% water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤0.5% single Impurity (redox titration)
≤1.0% mG1 (reversed phase HPLC, DMT-rG(ib)me-DMT)
≤3% residual Solvent content

color

white to off-white

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

compatibility

configured for MerMade

storage temp.

2-8°C

SMILES string

CO[C@@H]1[C@H](OP(OCCC#N)N(C(C)C)C(C)C)[C@@H](COC(c2ccccc2)(c3ccc(OC)cc3)c4ccc(OC)cc4)O[C@H]1n5cnc6C(=O)NC(NC(=O)C(C)C)=Nc56

InChI

1S/C45H56N7O9P/c1-28(2)41(53)49-44-48-40-37(42(54)50-44)47-27-51(40)43-39(57-9)38(61-62(59-25-13-24-46)52(29(3)4)30(5)6)36(60-43)26-58-45(31-14-11-10-12-15-31,32-16-20-34(55-7)21-17-32)33-18-22-35(56-8)23-19-33/h10-12,14-23,27-30,36,38-39,43H,13,25-26H2,1-9H3,(H2,48,49,50,53,54)/t36-,38-,39-,43-,62?/m1/s1

InChI key

IRRDHRZUOZNWDJ-MLLDKZSOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Proligo′s 2′O-Methyl RNA monomers are compatible with fast deprotection schemes that are based on the application of aliphatic amines, such as methylamine. The adenosine and guanosine monomers are protected with the standard benzoyl (bz) and isobutyryl (ib) groups.2′O-Methyl RNA is a nucleic acid analogue that is characterized by the exceptional hybridization properties that it imparts with complimentary DNA or RNA, as well as increased stability against enzymatic degradation compared to natural nucleic acids. The unique combination of properties of 2′O-Methyl RNA had found widespread use in the fields of:
  • Diagnostic probes
  • Aptamer and ribozyme development
  • Mixed 2′O-Methyl-RNA/DNA antisense molecule
Key Features:
  • High yield of crude oligonucleotides
  • Compatible with DNA synthesis
  • Can be employed together with DNA or RNA phosphoramidites in the same synthesis to produce mixmer oligonucleotides
  • Recommended deprotection conditions are 8 hours at 55 °C using concentrated ammonia solution, or with AMA (concentrated ammonia/40% aqueous methylamine I/I, v/v) for 10 minutes at 65 °C
  • Purification and other downstream processing of fully modified 2′OMethyl RNA oligonucleotides are simpler than in the case of RNA, as no special precautions are required to provide protection against nucleolyticdegradationDMT-2′O-Methyl-rG(ib) Phosphoramidite is configured for MerMade Synthesizers.

Other Notes

The synthesis cycle for 2′O-Methyloligoribonucleotides consists of the sameseries of reactions as the cycle that is employed for DNA monomers.However, the rate of coupling for 2′O-Methyl RNA monomers is slowercompared to that of DNA monomers (a coupling time of 6 minutes isrecommended for 2′O-Methyl RNA monomers compared to 90 seconds forDNA monomers). With the exception of the 2′O-Methyl RNA monomers andsupports, RNA synthesis is accomplished with the same reagents as DNAsynthesis. All 2′O-Methyl RNA phosphoramidites from Sigma-Aldrich arediluted with dry acetonitrile.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service