Skip to Content
Merck
All Photos(1)

Key Documents

G1627

Sigma-Aldrich

Gly-His

powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N4O3
CAS Number:
Molecular Weight:
212.21
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

Product Name

Gly-His,

form

powder

Quality Level

color

white

storage temp.

−20°C

SMILES string

NCC(=O)NC(Cc1cnc[nH]1)C(O)=O

InChI

1S/C8H12N4O3/c9-2-7(13)12-6(8(14)15)1-5-3-10-4-11-5/h3-4,6H,1-2,9H2,(H,10,11)(H,12,13)(H,14,15)

InChI key

YIWFXZNIBQBFHR-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Giulio Vistoli et al.
Biochemical and biophysical research communications, 492(3), 487-492 (2017-08-24)
The study combines HPLC-based with MS-based competitive analyses to evaluate the quenching activity of a set of carnosine derivatives towards methylglyoxal (MGO) and malondialdehyde (MDA) chosen as representative of α- and β-dicarbonyls, respectively. The obtained results underline that these derivatives
Javiera Cortés-Ríos et al.
Analytical biochemistry, 608, 113904-113904 (2020-08-18)
Amongst the available methodologies for protein determination, the bicinchoninic acid (BCA) assay highlights for its simplicity, sensitivity, repeatability and reproducibility. Nevertheless, in spite that the general principle behind this methodology is known, there are still unanswered questions regarding the chemistry
Urszula Rychlewska et al.
Dalton transactions (Cambridge, England : 2003), 39(38), 8906-8913 (2010-08-21)
Monocationic gold(III) complexes with histidine-containing peptides, glycyl-L-histidine (Gly-L-His) and L-alanyl-L-histidine (L-Ala-L-His) have been synthesized and characterized by (1)H NMR spectroscopy and X-ray crystallography. The crystallized Au(III) complexes, [Au(Gly-L-His-N,N',N'')Cl]NO(3)·1.25H(2)O and [Au(L-Ala-L-His-N,N',N'')Cl]NO(3)·2.5H(2)O, were obtained from water solution at pH < 1.0. The
N Manoj et al.
The journal of peptide research : official journal of the American Peptide Society, 56(4), 210-217 (2000-11-18)
The crystal structures of the complexes of oxalic acid with glycyl-L-histidine and L-histidyl-L-alanine were determined. The three crystallographically independent peptide molecules in the complexes have closed conformations. The terminal carboxyl group of the dipeptide and the oxalate ion in the
A Strauss et al.
FEMS microbiology letters, 127(3), 249-254 (1995-04-01)
A glycine-histidine tag (Gly3His6) was added to the C-terminus of a fusion protein consisting of the cholera toxin B-subunit (CtxB) and the IgA protease beta-domain (Iga beta). The aim was to facilitate single-step purification and to create a suitable tool

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service