Skip to Content
Merck
All Photos(1)

Key Documents

G0295

Sigma-Aldrich

Anti-GABAA Receptor (α6 subunit), Cytosolic Loop antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-γ-Aminobutyric Acid Type A Receptor (α6 subunit)

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€610.00

€610.00


Estimated to ship on28 May 2025



Select a Size

Change View
1 VIAL
€610.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€610.00


Estimated to ship on28 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

usage

sufficient for 10 blots

species reactivity

rat

technique(s)

western blot: 1:1000 using rat brain membrane fraction.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

γ -aminobutyric acid (GABA) receptor is the multifunctional, pentameric membrane proteins that operate GABA-gated chloride channels. It acts as major inhibitory neurotransmitter in the adult mammalian brain. It is the members of the neurotransmitter ligand-gated ion channels which plays various functions in the central nervous system, the peripheral nervous system, and in some nonneuronal tissues. At least four types of subunits (α, β, γ and δ) of GABA have been identified. Generally it contains one type of α and β subunit, and a single γ polypeptide in a ratio of 2:2:1.

Immunogen

fusion protein with the amino acid sequence representing the cytosolic loop of the rat GABAA receptor (α6 subunit)

Application

Anti-GABAA Receptor (α6 subunit), cytosolic loop antibody is suitable for western blot at a dilution of 1:1,000 using rat brain membrane fraction.

Biochem/physiol Actions

GABA is synthesized primarily from glutamate by glutamate decarboxylase (GAD). It is triggered by binding to its ligand-gated chloride channels, GABA(A) and GABA(C) as well as metabotropic receptor GABA(B). Along with neural development, it is also involved in a wide variety of physiological functions in tissues and organs outside the brain.

Target description

Detects 56-57 kDa GABAA receptor α6 subunit in rat brain membrane fractions.

Physical form

Solution in 10 mM HEPES, pH 7.5, 150 mM NaCl, 100 μg/ml BSA and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Giovannucci et al.
Nature neuroscience, 20(5), 727-734 (2017-03-21)
Cerebellar granule cells, which constitute half the brain's neurons, supply Purkinje cells with contextual information necessary for motor learning, but how they encode this information is unknown. Here we show, using two-photon microscopy to track neural activity over multiple days
U Rudolph et al.
Trends in pharmacological sciences, 22(4), 188-194 (2001-04-03)
The enhancement of GABA-mediated synaptic transmission underlies the pharmacotherapy of various neurological and psychiatric disorders. GABA(A) receptors are pluripotent drug targets that display an extraordinary structural heterogeneity: they are assembled from a repertoire of at least 18 subunits (alpha1-6, beta1-3
Richard W Olsen et al.
Neuropharmacology, 56(1), 141-148 (2008-09-02)
This mini-review attempts to update experimental evidence on the existence of GABA(A) receptor pharmacological subtypes and to produce a list of those native receptors that exist. GABA(A) receptors are chloride channels that mediate inhibitory neurotransmission. They are members of the
H Möhler et al.
Current opinion in pharmacology, 1(1), 22-25 (2001-11-20)
The GABA(A) receptor is a pluripotent drug target mediating anxiolytic, sedative, anticonvulsant, muscle relaxant and amnesic activity. These drug actions have now been attributed to defined receptor subtypes. Thus, precise guidelines are available for the development of novel drugs with
Structure and pharmacology of vertebrate GABAA receptor subtypes.
P J Whiting et al.
International review of neurobiology, 38, 95-138 (1995-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service