Skip to Content
Merck
All Photos(1)

Key Documents

FNCAS9TRACR

Sigma-Aldrich

SygRNA FnCas9 Synthetic tracrRNA

Sign Into View Organizational & Contract Pricing

Select a Size

5 NMOL
€83.50

€83.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 NMOL
€83.50

About This Item

UNSPSC Code:
41106609
NACRES:
NA.51

€83.50


Check Cart for Availability

Request a Bulk Order

packaging

pkg of 1 vial

shipped in

ambient

storage temp.

−20°C

General description

The SygRNA synthetic RNA system recruits FnCas9 nuclease to specific double stranded DNA targets. The FnCas9 tracrRNA (trans-activating crRNA) binds to a FnCas9 crRNA (CRISPR RNA) and the resulting crRNA-tracrRNA complex guides FnCas9 protein to crRNA-specified sites in double stranded DNA. SygRNA FnCas9 synthetic tracrRNA and crRNAs are optimized for use with FnCas9 protein and compatible with a variety of nucleic acid delivery and digestion methods. To purchase custom crRNAs click "Request Quotation for Custom crRNA Synthesis" at sigma.com/SygRNA

Application

Functional Genomics/Genome Editing/Molecular Biology/Molecular Cloning/Synthetic Biology

Features and Benefits

HPLC-purified SygRNA FnCas9 tracrRNA oligo is optimized for use with target-specific Francisella novicida Cas9 crRNA and FnCas9 protein for genome editing applications, large fragment molecular cloning, and programmable restriction digestion.

Physical form

Dry

Preparation Note

Resuspend SygRNA FnCas9 tracrRNA oligo in RNase-free Tris-containing buffer with a pH 8.0 or less.

Legal Information

SygRNA is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Combine guaranteed sgRNAs with our comprehensive range of CRISPR products and tools, including Cas9 and delivery reagents, for efficient genome modification with higher specificity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service