Skip to Content
Merck
All Photos(1)

Key Documents

FG4815

Sigma-Aldrich

DMEM/F-12

With stable glutamine, sodium pyruvate and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

DME/Nutrient Mixture F-12 Ham, DMEM Hams F12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71
Pricing and availability is not currently available.

Product Name

Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-alanyl-L-glutamine, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered
sterile

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, tested

components

phenol red: yes
stable glutamine: yes
glucose: high
HEPES: no
NaHCO3: yes
sodium pyruvate: yes

shipped in

ambient

storage temp.

2-8°C

General description

Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham is a 1:1 mixture. This DMEM F-12 Ham medium differs from the original formula because it is supplemented with L-alanyl-L-glutamine dipeptide. This dipeptide provides a more stable form of glutamine for use in cell cultures as glutamine alone is highly unstable in cell cultures.

Application

Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham has been used to culture human fibroblasts.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Nutrient Mixture F-12 Ham Formulation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service