Skip to Content
Merck

Skip To

F9932

FFN511 trifluoroacetate salt hydrate

≥98% (HPLC)

Synonym(s):

8-(2-Amino-ethyl)-2,3,5,6-tetrahydro-1H,4H-11-oxa-3a-aza-benzo[de]anthracen-10-one trifluoroacetate salt hydrate, 9-(2-aminoethyl)-2,3,6,7-tetrahydro-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizin-11-one trifluoroacetate salt hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Check Cart for Availability
€ 831,00

About This Item

Empirical Formula (Hill Notation):
C17H20N2O2 · F3CCO2H · xH2O
CAS Number:
Molecular Weight:
398.38 (anhydrous basis)
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated

€ 831,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

yellow

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

O.OC(=O)C(F)(F)F.NCCC1=CC(=O)Oc2c3CCCN4CCCc(cc12)c34

InChI

1S/C17H20N2O2.C2HF3O2.H2O/c18-6-5-11-10-15(20)21-17-13-4-2-8-19-7-1-3-12(16(13)19)9-14(11)17;3-2(4,5)1(6)7;/h9-10H,1-8,18H2;(H,6,7);1H2

InChI key

RRBOSSKRCWVUPA-UHFFFAOYSA-N

Biochem/physiol Actions

FFN511 is a fluorescent substrate for the synaptic vesicle monoamine transporters.
FFN511 is a fluorescent substrate for the synaptic vesicle monoamine transporters. FFN511 is used as an optical tracer of dopamine, a fluorescent false neurotransmitter (FFN) to visualize dopamine release from individual presynaptic terminals using time-lapse microscopsy fluorescence. The probe is sufficiently fluorescent so as to provide resolution of individual dopamine terminals at concentrations that do not interfere with normal catecholamine accumulation and transmission. FFN511 is compatible with GFPbased tags, the FM1-43 endocytic marker (Synapto Green), and other optical probes.

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1172P5749P186
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

solid

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage condition

desiccated

storage condition

desiccated

storage condition

-

storage condition

desiccated

solubility

DMSO: ≥20 mg/mL

solubility

DMSO: 10 mg/mL, clear

solubility

H2O: >5 mg/mL

solubility

H2O: 10 mg/mL, clear


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters