Skip to Content
Merck

Skip To

F9813

Fludarabine phosphate

Synonym(s):

2-Fluoro-9-(5-O-phosphono-β-D-arabinofuranosyl)-9H-purin-6-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H13FN5O7P
CAS Number:
Molecular Weight:
365.21
MDL number:
UNSPSC Code:
51102829
NACRES:
NA.85
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

color

white

solubility

DMSO: soluble

antibiotic activity spectrum

neoplastics

mode of action

DNA synthesis | interferes

storage temp.

−20°C

SMILES string

Fc1nc2[n](cnc2c(n1)[N+H3])C3OC(C(C3O)O)CO[P](=O)([O-])O

InChI

1S/C10H13FN5O7P/c11-10-14-7(12)4-8(15-10)16(2-13-4)9-6(18)5(17)3(23-9)1-22-24(19,20)21/h2-3,5-6,9,17-18H,1H2,(H2,12,14,15)(H2,19,20,21)

InChI key

GIUYCYHIANZCFB-UHFFFAOYSA-N

General description

Fludarabine is a purine analog included in the category of DNA-damaging drugs with well-known efficacy in B-cell chronic lymphocytic leukemia (B-CLL).[1]

Application


  • Characterization of Chemical Interactions between Clinical Drugs and the Oral Bacterium, Corynebacterium matruchotii, via Bioactivity-HiTES.: This study explores the interactions of clinical drugs like Fludarabine phosphate with Corynebacterium matruchotii, highlighting potential impacts on oral microbiota and implications for drug efficacy and safety (Lee DY et al., 2024).

  • Cocktail of lipophilic and hydrophilic chemotherapeutics in high-load core@shell nanocarriers to treat pancreatic tumours.: Investigates the efficacy of a combination of Fludarabine phosphate with other chemotherapeutics delivered via nanocarriers, aiming to enhance treatment outcomes for pancreatic cancer by improving drug delivery to the tumor site (Rudolph D et al., 2024).

  • Macrophage neogenin deficiency exacerbates myocardial remodeling and inflammation after acute myocardial infarction through JAK1-STAT1 signaling.: This research demonstrates the role of Fludarabine phosphate in modulating inflammation and cardiac repair post-myocardial infarction, offering insights into its potential therapeutic benefits beyond oncology (Zhang J et al., 2023).

  • SLC25A51 promotes tumor growth through sustaining mitochondria acetylation homeostasis and proline biogenesis.: Discusses the cellular mechanisms by which Fludarabine phosphate may influence metabolic pathways in cancer cells, highlighting its potential to disrupt tumor metabolism and promote cancer cell death (Li Y et al., 2023).

  • CD19-Targeting CAR T Cells for Myositis and Interstitial Lung Disease Associated With Antisynthetase Syndrome.: Reviews the use of Fludarabine phosphate in preconditioning regimens for CAR T-cell therapy, emphasizing its role in enhancing the efficacy of immunotherapy in treating autoimmune disorders (Pecher AC et al., 2023).


Biochem/physiol Actions

Fludarabine represses DNA replication and suppresses the nucleotide metabolism by inhibiting the enzyme ribonucleotide reductase.[2]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y0000419Y00022561272204
antibiotic activity spectrum

neoplastics

antibiotic activity spectrum

-

antibiotic activity spectrum

-

antibiotic activity spectrum

-

mode of action

DNA synthesis | interferes

mode of action

-

mode of action

-

mode of action

-

Quality Level

200

Quality Level

-

Quality Level

-

Quality Level

-

form

powder

form

-

form

-

form

-

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: soluble

solubility

-

solubility

-

solubility

-


Still not finding the right product?

Explore all of our products under Fludarabine phosphate


pictograms

Health hazard

signalword

Warning

Hazard Classifications

Muta. 2 - Repr. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. can i know how this product is delivered? In a sealed tube? or something else?

    1 answer
    1. This product is packaged in an amber glass bottle with a white polypropylene screw cap.

      Helpful?

Reviews

No rating value

Active Filters