Skip to Content
Merck
All Photos(1)

Key Documents

F9255

Sigma-Aldrich

Furaltadone hydrochloride

Synonym(s):

5-Morpholinomethyl-3-(5-nitrofurfurylideneamino)-2-oxazolidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N4O6 · HCl
CAS Number:
Molecular Weight:
360.75
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

Cl.O=C1OC(CN2CCOCC2)CN1\N=C\c3ccc(o3)N(=O)=O

InChI

1S/C13H16N4O6.ClH/c18-13-16(14-7-10-1-2-12(22-10)17(19)20)9-11(23-13)8-15-3-5-21-6-4-15;/h1-2,7,11H,3-6,8-9H2;1H/b14-7+;

InChI key

PPSVFZXMDMUIGB-FJUODKGNSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Leston et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(32), 3832-3836 (2011-11-23)
Presently, the rise of new contaminants in the environment has widened the scope of pharmaceutical analyses as to face the demanding new challenges. An increasing tendency for the interconnection and overlap of research fields, such as ecology and biochemistry, is
Peng Jie Luo et al.
Biomedical and environmental sciences : BES, 25(4), 449-457 (2012-10-03)
To determine 3-amino-5-morpholinomethyl-2-oxazolidinone (AMOZ) residues released from protein bound AMOZ in animal tissues. Polyclonal and monoclonal antibodies were produced in this study. A rapid, sensitive, and specific competitive direct enzyme-linked immunosorbent assay (cdELISA) was developed. Rabbit polyclonal antibodies were used
Jorge Barbosa et al.
Journal of agricultural and food chemistry, 60(17), 4227-4234 (2012-04-11)
The use of nitrofurans as veterinary drugs has been banned from intensive animal production in the European Union (EU) since 1993. The objective of the present study was to evaluate the accumulation and depletion of furaltadone and nifursol and their
A Stammati et al.
Cell biology and toxicology, 13(2), 125-130 (1997-02-01)
Nitrofuran drugs have been studied on cellular systems in order to develop in vitro tests for safety assessment of food contaminants. In the present study we have tested furaltadone on three cell lines (HEp-2, Caco-2 and V79), using the same
T Galeano Díaz et al.
Journal of chromatography. A, 764(2), 243-248 (1997-03-14)
A HPLC method with coulometric detection has been established to carry out the separation of the three nitrofuran derivatives, nitrofurantoin, furazolidone and furaltadone. A Nova-Pak C18 column (150 x 3.9 mm) and a Coulochem II detector from ESA have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service