Merck
Skip To
F8542
Nunc® TripleFlasks, Nunclon® Δ Surface
Culture flask, surface area 500 cm2, with solid cap, straight neck, sterile, case of 32 ea
Synonym(s):
cell culture flask, tissue culture flask, Nunc 132867, surface treated flask, tissue culture treated flask
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
1 case
Check Cart for Availability
€ 802,00
About This Item
NACRES:
NB.22
UNSPSC Code:
41122104
Surface area:
500 cm2 , 500 cm2 , culture area (cm2) per flask
Binding type:
Nunclon Delta surface
Material:
screw top blue high-density polyethylene cap, straight neck , straight neck transparent polystyrene flask
Sterility:
sterile
Packaging:
case of 32 ea
€ 802,00
Check Cart for Availability
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
screw top blue high-density polyethylene cap, straight neck , straight neck transparent polystyrene flask
sterility
sterile
packaging
case of 32 ea
manufacturer/tradename
Nunc 132867
flask working volume
200 mL
surface area
500 cm2 , 500 cm2 , culture area (cm2) per flask
working volume
200 mL
binding type
Nunclon Delta surface
General description
Perform cell culture expansion and experience 500cm2 total growth area within the footprint of a standard flask using the Nunc® TripleFlask™ Treated Cell Culture Flasks. Total culture area is achieved by way of the three parallel layers and a straight neck design that works effectively within automation systems. TripleFlask receives the Nunclon™ Delta treatment, a proprietary cell culture surface treatment that offers maximum adhesion for a broad range of cell types. Three layers of growth surfaces provide a total culture area of 500 cm2 in the same footprint as a standard Nunc 175cm2 flask.
Features and Benefits
  • Three parallel growth surfaces provide a total culture area of 500 cm2
  • External dimensions of a 175 cm2 standard flask
  • Ideal for scale-up cell culture applications
  • Extra caps in every carton
  • Nunclon™ Δ surface ensures consistent optimal cell attachment and growth
  • Barcoded with large Code 128
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
F8667TMO132920Z721905
binding type
Nunclon Delta surface
binding type
Nunclon Delta surface
binding type
Nunclon Delta surface
binding type
Nunclon Delta surface
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile; γ-irradiated
material
screw top blue high-density polyethylene cap, straight neck transparent polystyrene flask
material
screw top blue high-density polyethylene cap, straight neck , straight neck transparent polystyrene flask
material
screw top blue high-density polyethylene cap, straight neck , straight neck transparent polystyrene flask
material
canted neck transparent polystyrene flask, screw top blue high-density polyethylene cap (vent)
manufacturer/tradename
Nunc 132867
manufacturer/tradename
Nunc 132913
manufacturer/tradename
Nunc 132920
manufacturer/tradename
Nunc 159933
surface area
500 cm2 , 500 cm2 , culture area (cm2) per flask
surface area
500 cm2 , 500 cm2 , culture area (cm2) per flask
surface area
(500 cm2) , 500 cm2 , culture area (cm2) per flask
surface area
225 cm2
packaging
case of 32 ea
packaging
case of 32 ea
packaging
case of 32 ea
packaging
case of 30 ea (6 packs x 5 flasks)
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)