Skip to Content
Merck
All Photos(1)

Key Documents

F8305

Sigma-Aldrich

FRK (208-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

GTK, PTK5, RAK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

847-1147 nmol/min·mg

mol wt

~60 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FRK(2444)

General description

FRK (fyn-related kinase) is a tyrosine kinase, with a molecular weight of 54kDa. It contains SH2 and SH3 (src homology) domains. It is predominantly expressed in tissues of epithelial origin such as, liver, kidney, breast and colon. Its SH2 domain contains a putative bipartite nuclear localization signal.

Biochem/physiol Actions

FRK (fyn-related kinase) controls growth by phosphorylating perinuclear proteins. It inhibits cell growth both in a pRb (retinoblastoma)-dependent and independent manner. It cause cell cycle arrest at G1 phase in breast cancer cells. Certain mutations in this gene lead to its constitutive activation, as seen in hepatocellular adenomas. Its up-regulation in gliomas prevents both invasiveness and migration of glioma cells. This is attributable to its promotion of N-cadherin/β-catenin complex formation.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Camilla Pilati et al.
Cancer cell, 25(4), 428-441 (2014-04-17)
Hepatocellular adenomas (HCA) are benign liver tumors predominantly developed in women using oral contraceptives. Here, exome sequencing identified recurrent somatic FRK mutations that induce constitutive kinase activity, STAT3 activation, and cell proliferation sensitive to Src inhibitors. We also found uncommon
Qiong Shi et al.
Journal of molecular neuroscience : MN, 55(1), 32-41 (2014-06-28)
Fyn-related kinase (FRK), a member of Src-related tyrosine kinases, is recently reported to function as a potent tumor suppressor in several cancer types. Our previous study has also shown that FRK over-expression inhibited the migration and invasion of glioma cells.
Tanya Meyer et al.
International journal of cancer, 104(2), 139-146 (2003-02-06)
Rak is a 54 kDa protein tyrosine kinase originally isolated from breast cancer cells and expressed in epithelial cells. It resembles the protooncogene Src structurally but lacks an amino-terminal myristylation site and localizes to the nuclear and perinuclear regions of
Michael Welsh et al.
The Biochemical journal, 382(Pt 1), 261-268 (2004-06-10)
Hallmarks of the inflammatory process in Type I diabetes are macrophage activation, local release of beta-cell-toxic cytokines and infiltration of cytotoxic T lymphocytes. We have observed recently that mice overexpressing active FRK (fyn-related kinase)/RAK (previously named GTK/Bsk/IYK, where GTK stands
R J Craven et al.
Cancer research, 55(18), 3969-3972 (1995-09-15)
Rak is a nuclear tyrosine kinase containing Src homology 2 and 3 domains at its NH2 terminus. We report here that the retinoblastoma tumor susceptibility gene product pRb associates with Rak in vivo and in vitro. Rak binds in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service