Skip to Content
Merck

Skip To

F8065

Fucoidan from Macrocystis pyrifera

≥85%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mg
Check Cart for Availability
€ 156,00

About This Item

CAS Number:
MDL number:
NACRES:
NA.25
UNSPSC Code:
12352201

€ 156,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

algae (Macrocystis pyrifera)

Quality Segment

assay

≥85%

form

powder

color

white to brown

working range

4-7

solubility

water: 10 mg/mL, clear to hazy, colorless to very faintly brown

storage temp.

room temp

General description

Fucoidan is a fucose-rich, sulfated polysaccharide isolated from several species of brown seaweed.

Biochem/physiol Actions

Studies of fucoidan have found it to have a broad variety of biological activities including antitumor and antiangogenic activities, immunomodulatory, anti-inflammatory, anticoagulant, antithrombotic, and antioxidant activities. Fucoidan induces macrophage activation which subsequently activates mitogen activated protein kinases (MAPKs) and results in natural killer cell proliferation. Fucoidan acts as a nonselective selectin blocker and has been shown to induce apoptosis and suppress angiogenesis.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F8190F8315F5631
assay

≥85%

assay

≥95%

assay

≥95%

assay

-

biological source

algae (Macrocystis pyrifera)

biological source

algae (Fucus Vesiculosus)

biological source

algae (Undaria pinnatifida)

biological source

algae (Fucus Vesiculosus)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

solubility

water: 10 mg/mL, clear to hazy, colorless to very faintly brown

solubility

water: 10 mg/mL, clear, colorless to faintly yellow

solubility

water: 10 mg/mL, colorless to faintly yellow

solubility

H2O: 10 mg/mL, clear to hazy, faintly yellow to brownish-yellow

form

powder

form

powder

form

powder

form

powder

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters