Skip to Content
Merck
All Photos(1)

Key Documents

F7762

Sigma-Aldrich

Anti-Guinea Pig IgG (whole molecule) FITC antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

direct immunofluorescence: 1:64
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:320

storage temp.

−20°C

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Geffrotin et al.
Biochimica et biophysica acta, 1524(2-3), 196-202 (2000-12-13)
Interactions between tumour cells and surrounding extracellular matrix (ECM) influence the growth of tumour cells and their ability to metastasise. It is thus interesting to compare ECM composition in tumours and healthy tissues. Using the recently described MeLiM miniature pig
I G Makarenko et al.
Neuroscience letters, 328(2), 155-159 (2002-07-23)
This is the study first attempting to evaluate distribution of neurons expressing serotonin 5-hydroxytriptamine 1B (5-HT(1B)) receptors in hypothalamus by using immunocytochemistry. The 5-HT(1B)-immunoreactive neurons were widely distributed in hypothalamus. Accumulations of 5-HT(1B) neurons occurred in magnocellular nuclei, supraoptic nucleus
H Kasimiotis et al.
Diabetes, 49(4), 555-561 (2000-06-28)
The SOX (sex-determining region [SRY]-type high mobility group [HMG] box) family of transcription factors play key roles in determining cell fate during organ development. In this study, we have identified a new human SOX gene, SOX13, as encoding the type

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service