Skip to Content
Merck
All Photos(1)

Key Documents

F7301

Sigma-Aldrich

Fibroblast Growth Factor-18 human

>95% (SDS-PAGE and HPLC), recombinant, expressed in E. coli, lyophilized powder

Synonym(s):

FGF-18

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€859.00

€859.00


Estimated to ship on28 May 2025



Select a Size

Change View
25 μG
€859.00

About This Item

EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

€859.00


Estimated to ship on28 May 2025


biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

>95% (SDS-PAGE and HPLC)

form

lyophilized powder

potency

≤0.5 ng/mL

mol wt

predicted mol wt 21.2 kDa

packaging

pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... FGF18(8817)

General description

FGF18 (fibroblast growth factor 18) is a member of the FGF family, and is a part of the FGF8 subfamily, which also includes FGF8 and FGF17. It is a paracrine-acting protein, and does not undergo alternative splicing. It is produced as a precursor of 207 amino acids, from which the signal peptide of 27 hydrophobic amino acids is cleaved of.[1] This gene is localized to human chromosome 5q35.1, which produces a protein of 21.2kDa.[2]

Biochem/physiol Actions

FGF-18 is a heparin binding growth factor, which stimulates the proliferation and activation of cells that express the FGF receptors.
FGF18 (fibroblast growth factor 18) functions in both developing and adult organisms, and plays a predominant role in skeletal development. It is a putative early marker for prostate and ovarian cancer, and has potential as a target in the same.[1] It mediates gene profiling, and has anticatabolic effects mediated through Timp1 (tissue inhibitor of metalloproteinases). Thus, it has confers protection on adult articular cartilage.[3] In mice embryos, inactivation of this gene results in death due to aberrations in alveologenesis and skeleton development.[2]
Stimulates the proliferation and activation of cells that express the FGF receptors.

Physical form

Lyophilized from a 0.2 μm filtered solution in 10 mM Tris, pH 8.0 + 20mM NaCl.

Analysis Note

The biological activity is determined by the dose-dependent stimulation of thymidine uptake by BaF3 cells expressing FGF receptors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The crystal structure of fibroblast growth factor 18 (FGF18).
Alan Brown et al.
Protein & cell, 5(5), 343-347 (2014-03-29)
Yoshifumi Mori et al.
The Journal of biological chemistry, 289(14), 10192-10200 (2014-03-01)
To identify genes that maintain the homeostasis of adult articular cartilage and regenerate its lesions, we initially compared four types of chondrocytes: articular (AA) versus growth plate (AG) cartilage chondrocytes in adult rats, and superficial layer (IS) versus deep layer
Wei Wei et al.
The Journal of clinical investigation, 123(10), 4435-4448 (2013-09-11)
High-throughput genomic technologies have identified biomarkers and potential therapeutic targets for ovarian cancer. Comprehensive functional validation studies of the biological and clinical implications of these biomarkers are needed to advance them toward clinical use. Amplification of chromosomal region 5q31-5q35.3 has
Shuk Yee Ngan et al.
Current protocols, 2(1), e341-e341 (2022-01-14)
Human embryonic stem cells (ES) and induced pluripotent stem cells (iPSC) are powerful tools that have the potential to generate in vitro human lung epithelial cells. However, challenges in efficiency and reproducibility remain in utilizing the cells for therapy discovery
Amy P Wong et al.
Nature protocols, 10(3), 363-381 (2015-02-06)
Airway epithelial cells are of great interest for research on lung development, regeneration and disease modeling. This protocol describes how to generate cystic fibrosis (CF) transmembrane conductance regulator protein (CFTR)-expressing airway epithelial cells from human pluripotent stem cells (PSCs). The

Articles

Fibroblast growth factors in cell culture and various growth factors for your research

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service