Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C10H11FN2
CAS Number:
Molecular Weight:
178.21
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
eCl@ss:
32039090
assay
≥99%
storage temp.
−20°C
SMILES string
NCCc1c[nH]c2cc(F)ccc12
InChI
1S/C10H11FN2/c11-8-1-2-9-7(3-4-12)6-13-10(9)5-8/h1-2,5-6,13H,3-4,12H2
InChI key
BQTOKMYKZPCPRW-UHFFFAOYSA-N
Storage Class
13 - Non Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
PPE (personal protective equipment)
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.