Skip to Content
Merck
All Photos(1)

Key Documents

F7125

Sigma-Aldrich

6-Aminofluorescein

~95% (HPLC)

Synonym(s):

Fluoresceinamine isomer II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H13NO5
CAS Number:
Molecular Weight:
347.32
Beilstein:
51708
EC Number:
MDL number:
UNSPSC Code:
12171500

Assay

~95% (HPLC)

mp

285 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

Nc1ccc2C(=O)OC3(c4ccc(O)cc4Oc5cc(O)ccc35)c2c1

InChI

1S/C20H13NO5/c21-10-1-4-13-16(7-10)20(26-19(13)24)14-5-2-11(22)8-17(14)25-18-9-12(23)3-6-15(18)20/h1-9,22-23H,21H2

InChI key

YOAWSYSKQHLFPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

6-Aminofluorescein has been demonstrated to be useful fluorescent labeling reagent for fullerene-based liposome nanostructures termed ‘buckysomes′. 6-aminofluorescein dissolved in 0.1 N NaOH can be quantified from a maximum absorbance at 490 nm. Decarboxylated 6-aminofluorescein dissolved 6 N HCl (prepare by constant boiling at 110° C for 24 h) can be quantified from a maximum absorbance at 454 nm. Aldehyde groups are formed by oxidateive damage to proteins, which can be detected by conjugation to decarboxylated 6-aminofluorescein.

Other Notes

Note: Do not confuse with fluorescamine.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service