Skip to Content
Merck
All Photos(1)

Key Documents

F6301

Sigma-Aldrich

Fluorocurarine chloride

≥97% (TLC), from Vinca erecta, solid

Synonym(s):

Vincanine chlormethylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23ClN2O
CAS Number:
Molecular Weight:
342.86
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.77

biological source

Vinca erecta

Assay

≥97% (TLC)

form

solid

storage temp.

2-8°C

SMILES string

[Cl-].C\C=C1\C[N@+]2(C)CC[C@@]34[C@@H]2C[C@@H]1C(C=O)=C3Nc5ccccc45

InChI

1S/C20H22N2O.ClH/c1-3-13-11-22(2)9-8-20-16-6-4-5-7-17(16)21-19(20)15(12-23)14(13)10-18(20)22;/h3-7,12,14,18H,8-11H2,1-2H3;1H/b13-3-;/t14-,18-,20+,22-;/m0./s1

InChI key

CSLYOMBKQNZAED-CQLNDTFASA-N

Biochem/physiol Actions

Short-acting selective sympathetic ganglioblocker with weak antagonist activity on the nicotinic receptor at the neuromuscular junction; hypotensive.

Warning

Protect from light.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service