Skip to Content
Merck
All Photos(1)

Key Documents

F5882

Sigma-Aldrich

N-(3-[2-Furyl]acryloyl)-Ala-Lys

powder

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€239.00

€239.00


Estimated to ship on17 May 2025



Select a Size

Change View
50 MG
€239.00

About This Item

Empirical Formula (Hill Notation):
C16H23N3O5
CAS Number:
Molecular Weight:
337.37
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

€239.00


Estimated to ship on17 May 2025


Assay

≥98% (TLC)

Quality Level

form

powder

color

white to light yellow

solubility

water: 50 mg/mL
soluble, clear to slightly hazy

storage temp.

−20°C

SMILES string

C[C@H](NC(=O)\C=C\c1ccco1)C(=O)N[C@@H](CCCCN)C(O)=O

InChI

1S/C16H23N3O5/c1-11(18-14(20)8-7-12-5-4-10-24-12)15(21)19-13(16(22)23)6-2-3-9-17/h4-5,7-8,10-11,13H,2-3,6,9,17H2,1H3,(H,18,20)(H,19,21)(H,22,23)/b8-7+/t11-,13-/m0/s1

InChI key

BZUABSYGNBFITP-DDOQPMFUSA-N

Application

N-(3-[2-Furyl]acryloyl)-Ala-Lys has been used as a substrate in the carboxypeptidase B assay.

Substrates

Substrate for human plasma carboxypeptidase N.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B G Grimwood et al.
Analytical biochemistry, 170(1), 264-268 (1988-04-01)
A new high performance liquid chromatographic method has been developed for the determination of carboxypeptidase N activity which quantitates the furylacryloyl-alanine released by enzymatic cleavage of furylacryloyl-alanyl-lysine or furylacryloyl-alanyl-arginine. A short isocratic gradient elutes the substrate and product in less
Alejandra P Frasch et al.
Molecular and biochemical parasitology, 184(2), 63-70 (2012-05-12)
Metallocarboxypeptidases (MCP) of the M32 family of peptidases have been identified in a number of prokaryotic organisms but they are absent from eukaryotic genomes with the remarkable exception of those of trypanosomatids. The genome of Trypanosoma brucei, the causative agent
An improved spectrophotometric assay for human plasma carboxypeptidase N1.
T H Plummer et al.
Analytical biochemistry, 108(2), 348-353 (1980-11-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service