Skip to Content
Merck

F5304

Formalin, 10%, Neutral Buffered with 0.03% Eosin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shelf life

Expiry date on the label.

color

pink

pH

6.90-7.10

density

1.080 g/cm3

application(s)

hematology
histology

storage temp.

room temp

Application

Intended for use as general histological fixative. The addition of the eosin to the formulation allows the user to more easily visualize small fragments of tissue during the transfer of the tissue from the fixative to the tissue cassette.


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Muta. 2 - Skin Sens. 1

wgk

WGK 3

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

144.0 °F - closed cup

flash_point_c

62.2 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-56-1

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library