Skip to Content
Merck

F4640

3-Fluoro-1,2-propanediol

Synonym(s):

α-Fluorohydrin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
FCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
94.08
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


refractive index

n20/D 1.422 (lit.)

bp

102-103 °C/13 mmHg (lit.)

density

1.1 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OCC(O)CF

InChI

1S/C3H7FO2/c4-1-3(6)2-5/h3,5-6H,1-2H2

InChI key

PQDNJBQKAXAXBQ-UHFFFAOYSA-N



pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library