Skip to Content
Merck
All Photos(1)

Key Documents

F4303

Sigma-Aldrich

Fiduxosin hydrochloride

>98% (HPLC), solid

Synonym(s):

(3-[4-((3aR,9bR)-cis-9-methoxy-1,2,3,3a,4,9b-hexahydro-[1]-benzopyrano[3,4-c]Pyrrol-2-yl)butyl]-8-phenyl-pyrazino-[2′,3′:4,5]thieno [3,2-d]pyrimidine-2,4(1H,3H)-dione) hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H29N5O4S · HCl
CAS Number:
Molecular Weight:
592.11
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

>98% (HPLC)

form

solid

color

white

solubility

DMSO: soluble 10 mg/mL
H2O: insoluble

SMILES string

Cl[H].[H][C@@]12COc3cccc(OC)c3[C@]1([H])CN(CCCCN4C(=O)Nc5c(sc6ncc(nc56)-c7ccccc7)C4=O)C2

InChI

1S/C30H29N5O4S.ClH/c1-38-22-10-7-11-23-24(22)20-16-34(15-19(20)17-39-23)12-5-6-13-35-29(36)27-25(33-30(35)37)26-28(40-27)31-14-21(32-26)18-8-3-2-4-9-18;/h2-4,7-11,14,19-20H,5-6,12-13,15-17H2,1H3,(H,33,37);1H/t19-,20-;/m1./s1

InChI key

OFPMGRPQOZABPO-GZJHNZOKSA-N

Gene Information

Biochem/physiol Actions

α1-Adrenoceptor antagonist.

Features and Benefits

This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service