Skip to Content
Merck

Skip To

F3681

2-Furoyl-LIGRLO-amide trifluoroacetate salt

≥97% (HPLC)

Synonym(s):

2-Fly, 2-Furoyl-Leu-Ile-Gly-Arg-Leu-Orn-NH2 trifluoroacetate salt, 2-f-LIGRLO-NH2, 2-fAP, 2fLI, f-LIGRLO-NH2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
€ 169,00
€ 143,65

About This Item

Empirical Formula (Hill Notation):
C36H63N11O8 · xC2HF3O2
CAS Number:
Molecular Weight:
777.95 (free base basis)
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
51111800
MDL number:

€ 143,65

List Price€ 169,00Save 15%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

lyophilized powder

color

white

storage temp.

−20°C

SMILES string

Cc1cc(C)c2c(N)c(sc2n1)C(=O)NCc3ccc(Cl)cc3

InChI

1S/C17H16ClN3OS/c1-9-7-10(2)21-17-13(9)14(19)15(23-17)16(22)20-8-11-3-5-12(18)6-4-11/h3-7H,8,19H2,1-2H3,(H,20,22)

InChI key

FPRULFHDSFKYBV-UHFFFAOYSA-N

General description

2-Furoyl-LIGRLO-amide trifluoroacetate salt is a peptide that acts as a proteinase-activated receptor-2 (PAR2) agonist, and contains a furoyl group addition at its N-terminal.[1]

Application

2-Furoyl-LIGRLO-amide trifluoroacetate salt may be used as a protease-activated receptor 2 (PAR2) agonist in endothelial progenitor cells (EPCs) and in transient receptor potential cation channel subfamily V member (TRPV4)-transfected HEK293t cells.

Biochem/physiol Actions

2-Furoyl-LIGRLO-amide is a potent and selective protease-activated receptor 2 (PAR2) agonist.
2-Furoyl-LIGRLO-amide is a potent and selective protease-activated receptor 2 (PAR2) agonist. PAR-2 activation is associated with increases in cAMP and intracellular Ca(2+). Immunoblot analysis revealed increases in phosphorylation of epidermal growth factor (EGF) receptor (EGFR) tyrosine kinase, Src, Pyk2, cRaf, and ERK1/2 in response to PAR-2 activation. 2-Furoyl-LIGRLO-amide is nearly 100-fold more potent than SLIGRL-NH2 (Cat. No. S9317). 2-Furoyl-LIGRLO-amide caused both an endothelium-dependent relaxation and an endothelium-independent contraction. It produced delayed (6 hours later) facilitation of capsaicin-evoked visceral nociception, an effect being much more potent than SLIGRL-NH2. Such effects were mimicked by i.col. trypsin. 2-f-LIGRL-NH2, coadministered repeatedly with caerulein six times in total, abolished the caerulein-evoked abdominal hyperalgesia/allodynia in WT, but not PAR2-KO, mice. Repeated doses of 2-f-LIGRL-NH2 moderately attenuated the severity of caerulein-induced pancreatitis in WT animals. It induced a similar dose-dependent increase in Ca2 levels in the presence and absence of b-arrestins.
2-Furoyl-LIGRLO-amide trifluoroacetate salt is a highly potent agonist of proteinase-activated receptor-2 (PAR2).[1] In KNRK cells with high number of PAR2 receptors, this agonist causes elevation of intracellular Ca2+ levels. Studies on rat show that it causes the relaxation of aorta rings in an endothelium-dependent manner.[2]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S9317T7830SML2260
assay

≥97% (HPLC)

assay

≥97% (HPLC)

assay

>98% (HPLC)

assay

≥95% (HPLC)

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

-

form

lyophilized powder

form

solid

form

lyophilized powder

form

lyophilized powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

color

white

color

-

color

white to tan

color

white to beige


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters