Skip to Content
Merck

F3631

N-Formyl-Nle-Leu-Phe

≥97% (HPLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H33N3O5
CAS Number:
Molecular Weight:
419.51
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CCCCC(NC=O)C(=O)NC(CC(C)C)C(=O)NC(Cc1ccccc1)C(O)=O

Biochem/physiol Actions

Chemotactic peptide




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Structure and Biological Function.
Freer, R.J., et al.
Proceedings of the 6th American Peptide Symposium, 749-749 (1979)
P K Tsung et al.
Biochimica et biophysica acta, 541(2), 150-160 (1978-06-15)
1. Light- and heavy-plasma membrane fractions have been isolated from rabbit neutrophils and a chymotrypsin-like esterase has been shown to be present in these fractions. 2. The molecular weight of the chymotrypsin-like esterase of rabbit neutrophil plasma membrane was estimated