Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
2 mL | Check Cart for Availability | € 357,00 |
About This Item
biological source
sheep
Quality Segment
conjugate
FITC conjugate
antibody form
affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
form
buffered aqueous solution
technique(s)
direct immunofluorescence: 1:32
storage temp.
2-8°C
target post-translational modification
unmodified
General description
Application
Biochem/physiol Actions
Physical form
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) direct immunofluorescence: 1:32 | technique(s) direct immunofluorescence: 1:32 | technique(s) direct immunofluorescence: 1:128 | technique(s) direct immunofluorescence: 1:64 (minimum) |
| biological source sheep | biological source sheep | biological source sheep | biological source goat |
| conjugate FITC conjugate | conjugate FITC conjugate | conjugate FITC conjugate | conjugate FITC conjugate |
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| clone polyclonal | clone polyclonal | clone polyclonal | clone polyclonal |
Still not finding the right product?
Explore all of our products under Anti-Mouse IgG (whole molecule)–FITC antibody produced in sheep
— or —
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



