Skip to Content
Merck

Skip To

F2427

Fluprostenol isopropyl ester

≥98%, ethanol solution

Synonym(s):

(±)-16-(m-Trifluoromethylphenoxy)tetranorprostaglandin F2, Travoprost

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C26H35F3O6
CAS Number:
Molecular Weight:
500.55
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
200-578-6
MDL number:
Assay:
≥98%
Form:
ethanol solution
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98%

form

ethanol solution

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC(C)OC(=O)CCC\C=C/C[C@H]1[C@@H](O)C[C@@H](O)[C@@H]1\C=C\[C@@H](O)COc2cccc(c2)C(F)(F)F

InChI

1S/C26H35F3O6/c1-17(2)35-25(33)11-6-4-3-5-10-21-22(24(32)15-23(21)31)13-12-19(30)16-34-20-9-7-8-18(14-20)26(27,28)29/h3,5,7-9,12-14,17,19,21-24,30-32H,4,6,10-11,15-16H2,1-2H3/b5-3-,13-12+/t19-,21-,22-,23+,24-/m1/s1

InChI key

MKPLKVHSHYCHOC-AHTXBMBWSA-N

Gene Information

human ... PTGFR(5737)

Biochem/physiol Actions

Potent FP prostanoid receptor agonist used in animal models for glaucoma.

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2525P6617L0521
assay

≥98%

assay

≥98% (HPLC)

assay

≥97% (GC)

assay

-

form

ethanol solution

form

oil

form

ethanol solution

form

ethanol solution

Quality Level

200

Quality Level

-

Quality Level

200

Quality Level

200

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

wet ice

shipped in

-

shipped in

-

shipped in

dry ice

Gene Information

human ... PTGFR(5737)

Gene Information

-

Gene Information

-

Gene Information

-


pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

62.6 °F - closed cup

flash_point_c

17 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters