Skip to Content
Merck

F2153

Monoclonal Anti-Fibroblast Growth Factor-4 antibody produced in mouse

clone 19805.11, purified immunoglobulin, lyophilized powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
51111800
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

19805.11, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

indirect ELISA: suitable, neutralization: suitable, western blot: suitable

isotype

IgG1

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... FGF4(2249)

Immunogen

recombinant human fibroblast growth factor-4.

Biochem/physiol Actions

The antibody does not cross-react with bovine FGF-acidic, recombinant human FGF-acidic, bovine FGF-basic, recombinant human FGF-basic, and recombinant human β-ECGF.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Preparation Note

Purified using protein A


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

13 - Non Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library