EP6001082910
Eppendorf® Drawer style stainless steel rack for CryoCube® F740 upright freezers
closed configuration for storage of 20 boxes up to 136 x 136 x 63 mm (2.5 inch boxes); 5-Compartment, 5-Compartment, external W × D × H (140 mm) (563 mm) (339 mm)
About This Item
Recommended Products
material
stainless steel
feature
5-Compartment
Holds 20 x boxes (2.5 in.)
closed configuration for storage of 20 boxes up to 136 x 136 x 63 mm (2.5 inch boxes); 5-Compartment
manufacturer/tradename
Eppendorf® 6001082910
technique(s)
cryopreservation: suitable
external W × D × H
(140 mm) (563 mm) (339 mm)
size
563x140x344 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service