EP5948000210
Eppendorf® Centrifuge 5920 R G
AC/DC input 230 V AC, Schuko plug, incl. rotor S-4x-1000, high-capacity buckets and adapter 15/50 mL conical tubes
About This Item
Recommended Products
feature
CE compliant
refrigerated
to hold 108 15 mL conical tubes
to hold 4 high capacity buckets
to hold 52 50 mL conical tubes
incl. rotor S-4x-1000, high-capacity buckets and adapter 15/50 mL conical tubes
packaging
pkg of 1 ea
manufacturer/tradename
Eppendorf® 5948000210 (EU)
parameter
temp. range at -11-40 °C
13700 rpm
21194 × g
technique(s)
centrifugation: suitable
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Swing-bucket rotors and adapters accommodate tubes and bottles from 0.2 mL to 1,000 mL
- Plate rotor options for centrifugation of all types of MTP, PCR or Deepwell Plates
- Fixed-angle rotors for high-speed molecular biology applications in tubes from 0.2 mL to 50 mL
- High centrifugation speed of up to 21.194 × g (13,700 rpm)
- Aerosol-tight Eppendorf QuickLock caps and lids allow fast and ergonomic lid locking
- Centrifuge lid with soft-touch lid closure
- Quiet operation to improve your work environment
- Compact footprint saves valuable bench space
- Automatic rotor recognition and imbalance detection for maximum operational safety
- Temperature range from -11 °C to 40 °C
- FastTemp function for quick pre-cooling
- FastTemp pro function allows for automatic precooling based on pre-programmable time and date
- Continuous cooling maintains a constant temperature after the run has been completed – your samples stay cool
- ECO shut-off engages after 8 hours (adjustable) of non-use to reduce energy consumption and to extend compressor life
- Dynamic compressor control (DCC) technology for precise cooling performance
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service