EP5820756004
Eppendorf® Plate Removal Tool
used with rotor S-4-104, S-4x750
Synonym(s):
Eppendorf® Plate Removal Tool
About This Item
Recommended Products
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5820756004
technique(s)
centrifugation: suitable
compatibility
used with rotor S-4-104, S-4x750
General description
Application
Features and Benefits
2 pcs.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service