Key Documents
EP5810750008
Eppendorf® Adapter for 250 mL Rectangular Bucket
holds 25 x 1.2-5 mL tubes, set of 2 pieces
Select a Size
2 EA
€176.00
€176.00
Check Cart for Availability
Select a Size
Change View
2 EA
€176.00
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
€176.00
Check Cart for Availability
feature
holds 25 x 1.2-5 mL tubes
packaging
set of 2 pieces
manufacturer/tradename
Eppendorf® 5810750008
technique(s)
centrifugation: suitable
compatibility
for use with 250 mL reactangular bucket
General description
Tube adapter for Eppendorf 250 mL rectangular bucket.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)