Skip to Content
Merck
All Photos(1)

Key Documents

EP5810730007

Eppendorf® Rectangular buckets

size 500 mL, pkg of 4 ea

Synonym(s):

Lid for Eppendorf® FA-45-48-11 Rotor

Sign Into View Organizational & Contract Pricing

Select a Size

4 EA
€2,010.00

€2,010.00


Check Cart for Availability


Select a Size

Change View
4 EA
€2,010.00

About This Item

UNSPSC Code:
41103910
NACRES:
NB.82

€2,010.00


Check Cart for Availability

feature

for rotor A-4-81, S-4x500

packaging

pkg of 4 ea

manufacturer/tradename

Eppendorf® 5810730007

technique(s)

centrifugation: suitable

size

500 mL

Looking for similar products? Visit Product Comparison Guide

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service