EP5702732001
Eppendorf® Adapter for 85/100 mL Round Bucket
pack of 2 ea
Select a Size
€109.00
Select a Size
About This Item
€109.00
Recommended Products
feature
Holds 15 mL tubes
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5702732001
technique(s)
centrifugation: suitable
compatibility
for use with 100 mL round buckets
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service