EP5702711004
Eppendorf® Adapter for 90 mL rectangular bucket for use with standard tubes
Holds 9 mL tubes, pack of 2 ea
Select a Size
€105.00
Estimated to ship on22 May 2025
Select a Size
About This Item
€105.00
Estimated to ship on22 May 2025
Recommended Products
feature
Holds 9 mL tubes
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5702711004
technique(s)
centrifugation: suitable
compatibility
for use with 90 mL rectangular buckets
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service