Key Documents
EP5495512004
Rotor for Eppendorf® 5425 Centrifuge
S-96x0.2-PCR, to hold, 96 x 0.2 mL PCR tubes, to hold, 12 x 8-tube PCR strips, includes 4 x swing buckets
Select a Size
1 EA
€757.00
€757.00
Check Cart for Availability
Select a Size
Change View
1 EA
€757.00
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
€757.00
Check Cart for Availability
feature
to hold 12 x 8-tube PCR strips
to hold 96 x 0.2 mL PCR tubes
S-96x0.2-PCR
includes 4 x swing buckets
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5495512004
parameter
6,000 max. RCF (3,217 xg)
technique(s)
PCR: suitable
centrifugation: suitable
compatibility
for use with centrifuge models 5425
General description
Rotor for Eppendorf(R) 5425 Centrifuge
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)