EP5405000441
Eppendorf® Centrifuge 5425
knob control, with Rotor FA-24x2 including aerosol-tight QuickLock lid, AC/DC input 120 V AC, US 3-pin plug
Synonym(s):
Centrifuge 5425, centrifuge laboratory, lab centrifuge, micro centrifuge
About This Item
Recommended Products
feature
CSA approved
to hold 24 1.5/2.0 mL tubes
with Rotor FA-24x2 including aerosol-tight QuickLock lid
knob control
manufacturer/tradename
Eppendorf® 5405000441
parameter
15060 rpm speed (Max Speed )
technique(s)
centrifugation: suitable
capacity
10 x 5.0 mL(max rotor capacity)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
- Rotary knob control panel for easy parameter adjustments
- Equipped with Rotor FA-24x2 including aerosol-tight QuickLock lid
- Non-refrigerated
- 120 V, 50-60 Hz
- Has a main switch/power switch for switching the centrifuge on and off
- Reaches a maximum of 21,300 × g or 15,060 rpm
- Includes 3 program keys for easy selection of user-defined settings
- Offers over ten different acceleration and braking ramps for precise control
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service