Skip to Content
Merck
All Photos(1)

Key Documents

EP5308000003

Eppendorf® ThermoTop With Condens.Protect® Technology

for use with ThermoMixer F0.5/F1.5/F2.0/FP/C and ThermoStat C

Synonym(s):

Flow indicator

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€571.00

€571.00


Check Cart for Availability


Select a Size

Change View
1 EA
€571.00

About This Item

UNSPSC Code:
41102404
NACRES:
NB.77

€571.00


Check Cart for Availability

manufacturer/tradename

Eppendorf® 5308000003

Related Categories

General description

The Eppendorf® ThermoTop is developed by combining the features of the Eppendorf ThermoMixer and the heated lid approach of a polymerase chain reaction (PCR) cycler. The Eppendorf ThermoTop is a heatable lid equipped with condens.protect® technology. It not only prevents condensation but also enhances temperature homogeneity. With the Eppendorf ThermoTop, you can achieve optimal sample reaction conditions, leading to optimal results. Additionally, you can save time by eliminating the need for post-heating spinning of condensation-free tubes.

Features and Benefits

  • Ensure safe sample heating with efficient and reliable condensation prevention on tube lids.
  • Maintain constant reaction conditions for optimal enzymatic performance in your samples.
  • User-friendly handling with a wireless connection, automatic recognition, and operation.
  • Suitable for incubating light-sensitive samples, with or without heating function.
  • Simplify and streamline your workflow with the Eppendorf ThermoTop, eliminating the need for additional centrifugation steps to remove condensation droplets.
  • Ensure optimal sample protection with the Eppendorf ThermoTop, as its heated lid adjusts to the selected temperature on the instrument, preventing any adverse effects on sample temperature.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
condens.protect is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service