Skip to Content
Merck
All Photos(3)

Key Documents

EP4924000053

Eppendorf® Reference® 2 G

single-channel, variable, volume 10-100 μL, yellow, includes epT.I.P.S.® box

Synonym(s):

Adjustable volume pipette

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€384.00

€384.00


Check Cart for Availability


Select a Size

Change View
1 EA
€384.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

€384.00


Check Cart for Availability

feature

includes epT.I.P.S.® box
single-channel, variable

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 4924000053

volume

10-100 μL

color

yellow

Looking for similar products? Visit Product Comparison Guide

General description

Extraordinary precision and accuracy – Eppendorf´s premium pipette

Besides improved ergonomics the design focus lays on providing the most accurate results possible, optimum user safety, reliability and robustness for a long service life. The innovative single button operation is not only fast and easy, but it also allows active aerosol1) reduction and thus protects the user, the sample and the pipette. The sturdy handle manufactured with stainless steel protects the pipette from impact and in combination with the spring loaded bumpers the Reference 2 is likely to stay within calibration even if it takes a fall. These high levels of security make the Reference 2 ideal for use with precious liquids, or in any application requiring extremely high levels of accuracy. When there can be no compromise on quality and reproducibility, Eppendorf′s Reference 2 is an ideal instrument for these liquid handling applications.

Single-Button Operation

  • Lower Operating forces for aspiration/dispensing/tip ejection limit strain on the user.
  • Allows users to move more quickly through their pipetting tasks by not having to move the thumb from side to side to eject tips.
  • With the ability to eject a tip immediately following dispensing, aerosol contamination of your pipette is considerably reduced.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service