Skip to Content
Merck
All Photos(1)

Documents

EP4923601002

Sigma-Aldrich

Eppendorf® Reference 2 Pipettes with Carousel and epT.I.P.S.®

pipette carousel stand adapter, allows Reference® 2 Pipettors to be mounted on previous carousel stand versions

Synonym(s):

pipettor holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.02

material

plastic

feature

6 x mechanical pipette
allows Reference® 2 Pipettors to be mounted on previous carousel stand versions

manufacturer/tradename

Eppendorf® 4923601002

General description

Eppendorf Reference 2 Pipettes with Carousel and epT.I.P.S.

Application

Liquid handling

Features and Benefits

  • Mechanical air-cushion pipette with outstanding precision and accuracy - operated with one button
  • With its innovative one-button operation, the Reference 2 allows fast and ergonomic handling with reduced operating efforts and active aerosol reduction. A clear haptic feedback helps to avoid accidental tip ejection
  • Spring-loaded tip cones improve user-to-user reproducibility and ergonomics by lowering the tip attachment force
  • The unique smooth surface and full autoclavability guarantee efficient decontamination - making it the ideal companion when working under sterile conditions
  • A temporary adjustment allows for an easy adjustment of your Reference 2 for the most accurate pipetting of different liquids or in changing external conditions without the need for a full calibration
  • Available as single-channel pipette in fixed or variable volume as well as 8- and 12-channel pipette
  • The Reference 2 multi-channel variants offer high flexibility incl. a spring-loaded tip cone which can be switched on/off optionally

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service