Skip to Content
Merck
All Photos(1)

Key Documents

EP3125000133

Eppendorf Research® Plus Move It pipette

no. of channels, 8, volume 0.5-100 μL, 4.5-14 mm cone distance, light yellow

Synonym(s):

Eppendorf® Research® Plus Move It pipette, Pipettor, multichannel pipette, multichannel pipettor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01
Pricing and availability is not currently available.

material

light yellow

feature

adjustable
manual
no. of channels 8

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 3125000133

volume

0.5-100 μL

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf® Research plus Move It, 8-channel, 4.5-14 mm cone distance, variable, incl. epT.I.P.S.® Box 2.0, 5 – 100 µL, light yellow

Features and Benefits

  • 4-, 6-, 8- and 12-channel air-cushion pipettes with variable volume and adjustable tip spacing option
  • Available as mechanical (Research plus) and electronic (Xplorer plus) adjustable tip spacing multi-channel pipette
  • For plates (384-, 96-, 48-, 24- and 12-well), tubes, agarose gels and more
  • Tip spacing freely selectable from 4.5 to 33 mm, depending on pipette variant
  • Format limiter for quick switches backwards and forwards between source and destination formats
  • Manual adjustment knob for smooth vibration-free format switches and liquid transfers without dripping
  • Tubeless system for enhanced reliability and safety
  • Autoclavable for user and sample safety (Research plus fully, and Xplorer plus lower part only)
  • 360° rotatable pipette head for comfortable readability of display and fast identification of parameters
  • Optimally balanced pipette - reduces hand fatigue for stress-free operation

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Research is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service