Key Documents
EP3124000113
Eppendorf Research® plus Pipette, fixed volume
single-channel, volume 500 μL, blue
Select a Size
1 EA
€199.00
€199.00
Check Cart for Availability
Select a Size
Change View
1 EA
€199.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
€199.00
Check Cart for Availability
material
blue
feature
single-channel
mechanical
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 3121000112 (Global)
volume
500 μL
General description
The Eppendorf Research plus mechanical pipette is the result of over 60 years of experience in liquid handling and is one of the most frequently used pipettes in the world. Thanks to the groundbreaking Eppendorf PhysioCare Concept, the Research plus is very ergonomic and protects your health in the laboratory. It is characterized by low weight and low operating forces, is extremely reliable, fully autoclavable and easy to use. It feels good to know that you are doing the right thing – knowing that you are working with one of the most well-known and advanced pipettes in the world. A spring-loaded tip cone, an option for temporary adjustment, an improved volume display – and all this in a lightweight, fully autoclavable pipette: that is the Eppendorf Research plus manual pipette. In 2021, the Research plus single-channel variable volume pipettes were certified with the ACT Environmental Impact Factor Label by the non-profit organization My Green Lab . This makes them a good choice for laboratories looking for more sustainable products.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Research is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)