Key Documents
EP022639903
Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges
FA-45-6-30, aerosol-tight, to hold, 6 x 15/50 mL conical tubes, lid, including software upgrade (required for some older models)
About This Item
UNSPSC Code:
41103909
Pricing and availability is not currently available.
material
aluminum
feature
RCF 20,133 × g
to hold 6 x 15/50 mL conical tubes, lid, including software upgrade (required for some older models)
aerosol-tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022639903
compatibility
5804/5804R & 5810/5810R
FA-45-6-30
Related Categories
General description
Rotor FA-45-6-30, for 6 × 15/50 mL conical tubes, including software upgrade (required for some older models)
  • High-speed rotor for applications in tubes from 5 – 50 mL1)
  • Max. speed 20,133 × g (12,100 rpm) in Centrifuge 5810 R or 16,639 × g (11,000 rpm) in Centrifuge 5804/5804 R/58102)
  • Eppendorf QuickLock® rotor lid for fast lid locking
  • Aerosol-tight3) for safe centrifugation of hazardous samples
  • Low rotor weight (3.2 kg) for easy lifting
  • Anodized aluminum for high chemical resistance
  • 45° borehole angle minimizes pellets smear
  • Rotor, lid and adapters are autoclavable (121 °C, 20 min)
1) Please note that a software upgrade may be necessary for older centrifuges when using this rotor. Please contact your local Eppendorf organization or dealer for further information.
2) Rotors F-34-6-38 and FA-45-6-30 maintain 4 °C at 11,000 rpm
3) Aerosol-tightness tested by Public Health England, Porton Down, Great Britain
Features and Benefits
  • High-speed rotor for applications in tubes from 5 – 50 mL1)
  • Max. speed 20,133 × g (12,100 rpm) in Centrifuge 5810 R or 16,639 × g (11,000 rpm) in Centrifuge 5804/5804 R/58102)
  • Eppendorf QuickLock® rotor lid for fast lid locking
  • Aerosol-tight3) for safe centrifugation of hazardous samples
  • Low rotor weight (3.2 kg) for easy lifting
  • Anodized aluminum for high chemical resistance
  • 45° borehole angle minimizes pellets smear
  • Rotor, lid and adapters are autoclavable (121 °C, 20 min)
  • 1) Please note that a software upgrade may be necessary for older centrifuges when using this rotor. Please contact your local Eppendorf organization or dealer for further information.
  • 2) Rotors F-34-6-38 and FA-45-6-30 maintain 4 °C at 11,000 rpm
  • 3) Aerosol-tightness tested by Public Health England, Porton Down, Great Britain
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf QuickLock is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)