EP022492063
Eppendorf® epT.I.P.S.® standard
volume 50-1,250 μL, L 76 mm, green, colorless tip, pkg of 1000 ea (4 bags × 250 ea)
Synonym(s):
pipette tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
colorless tip
green
polypropylene
sterility
non-sterile
packaging
pkg of 1000 ea (4 bags × 250 ea)
manufacturer/tradename
Eppendorf® 022492063
L
76 mm
volume
50-1,250 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
epT.I.P.S.® Standard, Eppendorf Quality, 50 – 1,250 µL, 76 mm, green, colorless tips, 1,000 tips (4 bags × 250 tips)
- Original, high-quality Eppendorf pipette tips packaged in resealable bags
- Available in sizes from 10 µL to 10 mL in Eppendorf Quality purity grade
- epT.I.P.S. Standard 200 µL, 300 µL and 1,000 µL tips are also available color-coded yellow and blue
- epT.I.P.S. Standard pipette tips can be autoclaved at 121 °C in 20 min.
- Fine retracted or extended tips simplify work with small volumes
Features and Benefits
- Original, high-quality Eppendorf pipette tips packaged in resealable bags
- Available in sizes from 10 µL to 10 mL in Eppendorf Quality purity grade
- epT.I.P.S. Standard 200 µL, 300 µL and 1,000 µL tips are also available color-coded yellow and blue
- epT.I.P.S. Standard pipette tips can be autoclaved at 121 °C in 20 min.
- Fine retracted or extended tips simplify work with small volumes
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service