Skip to Content
Merck
All Photos(1)

Documents

EP022431048

Eppendorf® DNA LoBind tubes

capacity 2.0 mL, PCR clean, pkg of 250 ea (5 x 50ea)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103037

material

cap (push fit)
conical bottom
polypropylene
polypropylene cap

sterility

non-sterile

feature

DNase free
PCR clean
RCF 20,000 × g
RNase free
graduations

packaging

pkg of 250 ea (5 x 50ea)

manufacturer/tradename

Eppendorf® 022431048

capacity

2.0 mL

diam.

10.5 mm

color

clear

suitability

suitable for PCR

binding type

low binding surface

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

DNA LoBind® Tubes, snap cap, DNA LoBind®, 2.0 mL, PCR clean, colorless, 250 tubes (5 bags × 50 tubes)
  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • Precise lid sealing for minimal evaporation rates in tube format
  • Rated up to 30,000_x_g (25,000 × g for 2,0 mL) centrifugation speed for molecular biology applications

Features and Benefits

Signifcantly reduce sample-to-surface binding to ensure maximal recovery of DNA and RNA molecules

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Eppendorf LoBind is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nikos Tsolakos et al.
Vaccine, 28(18), 3211-3218 (2010-03-02)
In this study, we evaluated the effect of the growth medium on the composition and immunogenicity of meningococcal outer membrane vesicle (OMV) vaccines after cultivation of the Norwegian serogroup B 44/76 vaccine strain in either Frantz' or modified Catlin-6 media
Sharon N Finger et al.
Nucleic acids research, 36(4), 1260-1272 (2008-01-05)
Telomerase is a ribonucleoprotein enzyme that maintains chromosome ends through de novo addition of telomeric DNA. The ability of telomerase to interact with its DNA substrate at sites outside its catalytic centre ('anchor sites') is important for its unique ability
E I Trilisky et al.
Journal of chromatography. A, 1142(1), 2-12 (2007-01-24)
Purified viruses are used in gene therapy and vaccine production. Ion-exchange chromatography (IEC) is the most common method for large-scale downstream purification of viruses and proteins. Published IEC protocols provide details for specific separations but not general methods for selecting
M Yang et al.
International journal of pharmaceutics, 331(2), 176-181 (2006-11-28)
Salmon calcitonin (sCT) powders suitable for inhalation, containing chitosan and mannitol as absorption enhancer and protection agent, respectively, were prepared using a spray-drying process. The effect of chitosan on physicochemical stability of sCT in the dry powder was investigated by
Thandavarayan Kathiresan et al.
Methods in molecular biology (Clifton, N.J.), 493, 269-286 (2008-10-08)
Functional proteomics comprises a wide range of technologies for the identification of novel protein-protein interactions and biological markers. Studies of protein-protein interactions have gained from the development of techniques and technologies such as immunoprecipitation, preparative two-dimensional (2-D) gel electrophoresis for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service