EP0030524101
Eppendorf® Deepwell plates, Protein LoBind, 384 wells
white plate, conical bottom colorless wells, capacity 200 μL, pkg of 40 ea
Select a Size
€886.00
Estimated to ship on21 May 2025
Select a Size
About This Item
white plate
€886.00
Estimated to ship on21 May 2025
Recommended Products
material
conical bottom colorless wells
white plate
packaging
pkg of 40 ea
manufacturer/tradename
Eppendorf® 0030524101
technique(s)
PCR: suitable
capacity
200 μL
size
384 wells
working volume
200 μL
Looking for similar products? Visit Product Comparison Guide
General description
- Eppendorf® Deepwell plates, Protein LoBind are available as PCR clean purity grade, free of human DNA, DNase, RNase and PCR inhibitors clean. They are used to maximize sample recovery by significantly reducing sample binding to the surface.
- They comprise of a two-component polymer mix which creates a hydrophilic surface that ensures optimized recovery rates of valuable samples.
- Free of surface coating (e.g., silicone) to minimize the risk of sample interference.
- Available in tube, microplate, and deepwell plate formats for easy-up scaling.
- High-contrast Unique OptiTrack® matrix: up to 30% faster sample identification and fewer pipetting errors.
- RecoverMax® well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
- Raised well rims and a smooth surface guarantee reliable sealing in plates.
Application
- Preparation or storage of protein, peptide or antibody samples
- Enzymatic assays – the hydrophilic surface reduces denaturation when interacting with the vessel wall
- Avoiding sample loss when storing virus stock solutions
- Storage of cell suspensions
- Sample storage and preparation
- Bacteria and yeast cultivation.
- DNA and RNA isolation with high throughput.
- Storage of cDNA or genomic banks.
- Storage of siRNA or oligonucleotide libraries.
- Protein analysis.
- Storage of valuable protein and DNA samples in Eppendorf LoBind plates.
- Plasmid purification with high throughput.
- Creating dilution series.
Features and Benefits
- Eppendorf LoBind material ensures outstanding sample recovery for better test results
- Without surface coating (e.g. silicone) to minimize the risk of sample interference
- Batch-certified purity grade PCR clean: free from human DNA, DNase, RNase and PCR inhibitors
- Available as reaction vessel, microplate and deepwell plate for easy scaling
- Unique high-contrast OptiTrack matrix: up to 30% faster sample detection and fewer pipetting errors
- RecoverMax well design: Optimized well geometry for minimized residual or dead volume and excellent mixing properties
- Raised corrugated edges and a smooth surface ensure reliable sealing of the plates
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service