Key Documents
EP0030129334
Eppendorf® Twin tec microbiology PCR plates, 96 wells
blue plate, 96, semi-skirted, conical bottom colorless wells, semi-skirted, PCR clean, pkg of 10 ea, blue
About This Item
UNSPSC Code:
41121800
NACRES:
NC.22
material:
blue plate
conical bottom colorless wells
size:
96 wells
feature:
PCR clean
skirt
96, semi-skirted
material
blue plate
conical bottom colorless wells
feature
PCR clean
skirt
96, semi-skirted
packaging
pkg of 10 ea
manufacturer/tradename
Eppendorf® 0030129334
size
96 wells
color
blue
Features and Benefits
  • Individually blistered Certified PCR clean
  • Additionally free of bacterial DNA
  • For all applications that need ultimate purity or certified absense of bacterial DNA
  • Ideal for quantitative real-time PCR as well – OptiTrack matrix for faster sample identification and fewer pipetting errors
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)